Honeywell™ North™ Silver Shield™ Chemical Resistant Apparel
Various chemical resistant apparel
Manufacturer:  Honeywell™ SSCAXL
 View more versions of this product
Catalog No. 19-088-1683D
Description
Description
  • Five layer PE and EVOH constructions provides excellent chemical resistant to over 280 different chemicals including alchohols, aliphatics, aromatice, chlorines, ketones and esters
  • Provides high performance protection in the most demanding applications
Specifications
Specifications
Silver
X-Large
Composite Fabric
Coat Apron (with sleeves)