Honeywell™ C-Safe Circuit Breaker Lockouts
Manufacturer:  Honeywell™ CB02
 View more versions of this product
Catalog No. 29-664-190
Description
Description
Specifications
Specifications
2.62 x 1 x 1.8 in. (66 x 25.4 x 48mm)
1.8 in.
0.3 in. hole
66mm
Double
25.4mm
Polyamide Plastic
Manufacturing, mining, municipal services, oil and gas, utilities
48mm
2.62 in.
223°C
1 in.
Red
Circuit Breaker Lockout