Honeywell™ B-Safe Lockout/Tagout Devices for Valves
Manufacturer:  Honeywell™ BS02R
 View more versions of this product
Catalog No. 29-664-186
Description
Description
Specifications
Specifications
Red
0.78cm
1.5 to 2.5 in ball valve
-46° to 182°C
Lockout/Tagout Device
0.31 in.
Locking out quarter turn ball valves with flat 'dog-leg' handles
-50° to 360°F
Polypropylene