Honeywell™ B-Safe Lockout/Tagout Devices for Valves
Manufacturer:  Honeywell™ BS01R
 View more versions of this product
Catalog No. 29-664-185
Description
Description
Specifications
Specifications
Red
2 to 8 in. ball valve
-46° to 182°C
2cm
Lockout/Tagout Device
To prevent accidental activation
-50° to 360°F
0.87 in.
Polypropylene