Honeywell™ B-Safe Lockout/Tagout Devices for Valves
Manufacturer:  Honeywell™ BS04R
 View more versions of this product
Catalog No. 29-664-188
Description
Description
Specifications
Specifications
Red
0.93cm
Butterfly valve
-46° to 182°C
4cm
Lockout/Tagout Device
0.37 in.
Manufacturing, mining, municipal services, oil and gas, utilities
-50° to 360°F
1.62 in.
Polypropylene