Honeywell B-Safe Lockout/Tagout Devices for Valves
$17.58 - $44.88
Specifications
Material Polypropylene
Product Type Lockout/Tagout Device
Temperature Range (English) -50° to 360°F
Temperature Range (Metric) -46° to 182°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color For Use With (Application) For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Color For Use With (Application) For Use With (Equipment) Price Quantity  
29-664-185 Honeywell
BS01R
Red To prevent accidental activation 2 to 8 in. ball valve Each for $17.58
29-664-186 Honeywell
BS02R
Red Locking out quarter turn ball valves with flat 'dog-leg' handles 1.5 to 2.5 in ball valve Each for $28.43
29-664-187 Honeywell
BS03R
Red Manufacturing, mining, municipal services, oil and gas, utilities 2 to 8 in. ball valve Each for $34.07
19-170-193 Honeywell
BS04B
Blue Manufacturing, mining, municipal services, oil and gas, utilities Butterfly valve Each for $44.88
29-664-188 Honeywell
BS04R
Red Manufacturing, mining, municipal services, oil and gas, utilities Butterfly valve Each for $44.75
Specifications Description & Specifications
Specifications
Polypropylene
Lockout/Tagout Device
-50° to 360°F
-46° to 182°C