missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Homoveratrylamine 98.0+%, TCI America™

$59.05 - $619.25

Chemical Identifiers

CAS 120-20-7
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.24
MDL Number MFCD00008188
InChI Key ANOUKFYBOAKOIR-UHFFFAOYSA-N
Synonym 2-3,4-dimethoxyphenyl ethanamine, 2-3,4-dimethoxyphenyl ethylamine, 3,4-dimethoxyphenethylamine, 3,4-dimethoxyphenylethylamine, dimethoxydopamine, dimethoxyphenylethylamine, dimethylmescaline, dimpea, dmpea, homoveratrylamine
PubChem CID 8421
ChEBI CHEBI:136995
IUPAC Name 2-(3,4-dimethoxyphenyl)ethan-1-amine
SMILES COC1=CC=C(CCN)C=C1OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D067825G
View Documents Promotion Details
Tci America
D067825G
25g
Each for $59.05
 
D0678500G
View Documents Promotion Details
Tci America
D0678500G
500g
Each for $619.25
 
Description

Description

Specifications

Chemical Identifiers

120-20-7
181.24
ANOUKFYBOAKOIR-UHFFFAOYSA-N
8421
2-(3,4-dimethoxyphenyl)ethan-1-amine
C10H15NO2
MFCD00008188
2-3,4-dimethoxyphenyl ethanamine, 2-3,4-dimethoxyphenyl ethylamine, 3,4-dimethoxyphenethylamine, 3,4-dimethoxyphenylethylamine, dimethoxydopamine, dimethoxyphenylethylamine, dimethylmescaline, dimpea, dmpea, homoveratrylamine
CHEBI:136995
COC1=CC=C(CCN)C=C1OC

Specifications

156°C
25g
C10H15NO2
2735
ANOUKFYBOAKOIR-UHFFFAOYSA-N
2-(3,4-dimethoxyphenyl)ethan-1-amine
8421
181.24
≥98.0% (GC,T)
Undesignated
120-20-7
MFCD00008188
2-3,4-dimethoxyphenyl ethanamine, 2-3,4-dimethoxyphenyl ethylamine, 3,4-dimethoxyphenethylamine, 3,4-dimethoxyphenylethylamine, dimethoxydopamine, dimethoxyphenylethylamine, dimethylmescaline, dimpea, dmpea, homoveratrylamine
COC1=CC=C(CCN)C=C1OC
181.24
CHEBI:136995
Clear Liquid at 20°C
Homoveratrylamine
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0742

RTECSNumber : SH2300000

TSCA : Yes

SDS
Product Certifications