missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Homovanillic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 162530050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Homovanillic acid | |
306-08-1 | |
97.5% min. (HPLC) | |
C9H10O4 | |
MFCD00004350 | |
10, 409 | |
homovanillic acid, 4-hydroxy-3-methoxyphenylacetic acid, 2-4-hydroxy-3-methoxyphenyl acetic acid, vanillacetic acid, benzeneacetic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxyphenylacetic acid, homovaniuic acid, 4-hydroxy-3-methoxyphenyl acetic acid, homovanillate, 4-hydroxy-3-methoxybenzeneacetic acid | |
QRMZSPFSDQBLIX-UHFFFAOYSA-N | |
2-(4-hydroxy-3-methoxyphenyl)acetic acid | |
1738 | |
182.18 |
98% | |
Authentic | |
Glass bottle | |
HOC6H3(OCH3)CH2CO2H | |
5g | |
15, 4779 | |
Solubility in water: soluble. Other solubilities: slightly soluble in alcohol,ether,soluble in benzene,insoluble in cyclohexane | |
COC1=C(C=CC(=C1)CC(=O)O)O | |
182.18 | |
CHEBI:545959 | |
98% |
Safety and Handling
AIR SENSITIVE
EINECSNumber : 206-176-7
TSCA : TSCA
RUO â Research Use Only