missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Homovanillic acid, 98+%

Manufacturer:  Alfa Aesar™ A1244103

 View more versions of this product

Catalog No. AAA1244103



Description

Description

Solubility
Soluble in water,dimethyl sulfoxide, ethanol, methanol and ethyl acetate.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Homovanillic acid
1g
C9H10O4
2213447
homovanillic acid, 4-hydroxy-3-methoxyphenylacetic acid, 2-4-hydroxy-3-methoxyphenyl acetic acid, vanillacetic acid, benzeneacetic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxyphenylacetic acid, homovaniuic acid, 4-hydroxy-3-methoxyphenyl acetic acid, homovanillate, 4-hydroxy-3-methoxybenzeneacetic acid
COC1=C(C=CC(=C1)CC(=O)O)O
182.175
CHEBI:545959
≥98%
140°C to 145°C
306-08-1
MFCD00004350
14,4740
QRMZSPFSDQBLIX-UHFFFAOYSA-N
2-(4-hydroxy-3-methoxyphenyl)acetic acid
1738
182.18

Chemical Identifiers

306-08-1
182.175
QRMZSPFSDQBLIX-UHFFFAOYSA-N
1738
2-(4-hydroxy-3-methoxyphenyl)acetic acid
C9H10O4
MFCD00004350
homovanillic acid, 4-hydroxy-3-methoxyphenylacetic acid, 2-4-hydroxy-3-methoxyphenyl acetic acid, vanillacetic acid, benzeneacetic acid, 4-hydroxy-3-methoxy, 3-methoxy-4-hydroxyphenylacetic acid, homovaniuic acid, 4-hydroxy-3-methoxyphenyl acetic acid, homovanillate, 4-hydroxy-3-methoxybenzeneacetic acid
CHEBI:545959
COC1=C(C=CC(=C1)CC(=O)O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents