Please login to your online account to display your discounted pricing

Homoquinolinic acid, Tocris

Selective, potent NMDA agonist

$238.81 - $984.19

Chemical Identifiers

Chemical Name or Material Homoquinolinic acid
Assay Percent Range >99%
Molecular Formula C8H7NO4
Formula Weight 181.15
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

019710

 
r & d systems
RDS019710
10mg Each for $238.81

019750

 
r & d systems
RDS019750
50mg Each for $984.19
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 181.15
Chemical Name or Material Homoquinolinic acid
Assay Percent Range >99%
Molecular Formula C8H7NO4
CAS 490-75-5
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature