Please login to your online account to display your discounted pricing

Homoquinolinic acid, Tocris


Selective, potent NMDA agonist

Quantity 50mg

Manufacturer: r & d systems  RDS019750

 View more versions of this product

Catalog No. 019750

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 181.15
Chemical Name or Material Homoquinolinic acid
Assay Percent Range >99%
Molecular Formula C8H7NO4
CAS 490-75-5
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Homoquinolinic acid
Assay Percent Range >99%
Molecular Formula C8H7NO4
Formula Weight 181.15
Accessories

Store at Room Temperature