Please login to your online account to display your discounted pricing

Homopiperazine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Homopiperazine
CAS 505-66-8
Assay 97.5 to 102.5% (on dry substance)
Assay Percent Range 98%
Molecular Formula C5H12N2
Formula Weight 100.16
MDL Number MFCD00006933
Synonym 1, 4-Diazacycloheptane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC120851000

  SDS
thermo fisher scientific
120851000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC120855000

  SDS
thermo fisher scientific
AC120855000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Mass
Formula Weight 100.16
Quantity 100g
Color White to light yellow
Assay 97.5 to 102.5% (on dry substance)
Melting Point 38° to 42°C
Chemical Name or Material Homopiperazine
Water 1% max.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C5H12N2
Packaging Glass bottle
MDL Number MFCD00006933
CAS 505-66-8
Synonym 1, 4-Diazacycloheptane
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.