missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Homopiperazine, 98%

Manufacturer:  Alfa Aesar™ A1575136

 View more versions of this product

Catalog No. AAA1575136



Description

Description

Applications
Homopiperazine is used to prepare 1,4-bis-(2-thiazolyl)-1,4-diazacycloheptane by reacting with 2-bromo-thiazole. It acts as an intermediate for repaglinide and fasudil hydrochloride. It is also used in the preparation of potent H3 receptor antagonists, which is used for the treatment of neurodegenerative conditions. Further, it is used in vesicant, cosmetic, emulsifier and energetic material. It acts as a corrosion inhibitor for iron. In pharmaceuticals, it is employed as antipyretic and reducing blood sugar, adiposity, calm, antipsychotic and anti-nervous.

Solubility
Soluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

Homopiperazine
Amine-like
38°C to 44°C
505-66-8
MFCD00006933
102711
FQUYSHZXSKYCSY-UHFFFAOYSA-N
1,4-diazepane
68163
98%
64°C (147°F)
168°C to 170°C
500g
C5H12N2
UN3259
homopiperazine, 1,4-diazacycloheptane, 1h-1,4-diazepine, hexahydro, hexahydro-1,4-diazepine, 1,4 diazepane, perhydro-1,4-diazepine, trimethyleneethylenediamine, 1,4-diazepan, unii-95cl167w8t, hexahydro-1h-1,4-diazepine
C1CNCCNC1
100.165
100.17

Chemical Identifiers

505-66-8
100.165
FQUYSHZXSKYCSY-UHFFFAOYSA-N
68163
C1CNCCNC1
C5H12N2
MFCD00006933
homopiperazine, 1,4-diazacycloheptane, 1h-1,4-diazepine, hexahydro, hexahydro-1,4-diazepine, 1,4 diazepane, perhydro-1,4-diazepine, trimethyleneethylenediamine, 1,4-diazepan, unii-95cl167w8t, hexahydro-1h-1,4-diazepine
1,4-diazepane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

RTECSNumber : HM3850000

TSCA : TSCA

Documents