missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Homophthalonitrile, tech. 90%

Manufacturer:  Alfa Aesar™ L1241914

 View more versions of this product

Catalog No. AAL1241914



Specifications

Specifications

Homophthalonitrile
25g
C9H6N2
UN3439
2-cyanomethyl benzonitrile, homophthalonitrile, 2-cyanobenzyl cyanide, 2-cyanophenylacetonitrile, alpha-cyano-o-tolunitrile, o-cyanobenzyl cyanide, benzeneacetonitrile, 2-cyano, 2-cyanobenzeneacetonitrile, o-cyanophenyl acetonitrile, o-tolunitrile, alpha-cyano
C1=CC=C(C(=C1)CC#N)C#N
142.161
142.16
Technical
73°C to 79°C
3759-28-2
MFCD00001895
1940497
GKHSEDFDYXZGCG-UHFFFAOYSA-N
2-(cyanomethyl)benzonitrile
77368
90%

Chemical Identifiers

3759-28-2
142.161
GKHSEDFDYXZGCG-UHFFFAOYSA-N
77368
C1=CC=C(C(=C1)CC#N)C#N
C9H6N2
MFCD00001895
2-cyanomethyl benzonitrile, homophthalonitrile, 2-cyanobenzyl cyanide, 2-cyanophenylacetonitrile, alpha-cyano-o-tolunitrile, o-cyanobenzyl cyanide, benzeneacetonitrile, 2-cyano, 2-cyanobenzeneacetonitrile, o-cyanophenyl acetonitrile, o-tolunitrile, alpha-cyano
2-(cyanomethyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

RTECSNumber : XV0930000

Documents