Learn More
Homophthalic anhydride, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1553830
Description
It is used as a chemical and organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a chemical and organic intermediates.
Solubility
Decomposes in water.
Notes
Moisture sensitive. Storage temperature 15 - 30°C. Store under inert gas. Store away from exposure to moisture, strong oxidizing agents, strong acids, strong bases.
Specifications
Homophthalic anhydride | |
140°C to 144°C | |
MFCD00006894 | |
5244 | |
homophthalic anhydride, 1,3-isochromandione, isochroman-1,3-dione, 1h-2-benzopyran-1,3 4h-dione, homophthalic acid anhydride, 4h-2-benzopyran-1,3-dione, 1h-isochromene-1,3 4h-dione, 3,4-dihydro-1h-2-benzopyran-1,3-dione, 4h-benzo c pyran-1,3-dione, homophthalic anhydride 2-carboxyphenylacetic anhydride | |
AKHSBAVQPIRVAG-UHFFFAOYSA-N | |
4H-isochromene-1,3-dione | |
12801 | |
≥97% |
703-59-3 | |
C9H6O3 | |
250 g | |
Moisture sensitive | |
Decomposes in water. | |
C1C2=CC=CC=C2C(=O)OC1=O | |
162.144 | |
162.14 |
Chemical Identifiers
703-59-3 | |
162.144 | |
AKHSBAVQPIRVAG-UHFFFAOYSA-N | |
12801 | |
C1C2=CC=CC=C2C(=O)OC1=O |
C9H6O3 | |
MFCD00006894 | |
homophthalic anhydride, 1,3-isochromandione, isochroman-1,3-dione, 1h-2-benzopyran-1,3 4h-dione, homophthalic acid anhydride, 4h-2-benzopyran-1,3-dione, 1h-isochromene-1,3 4h-dione, 3,4-dihydro-1h-2-benzopyran-1,3-dione, 4h-benzo c pyran-1,3-dione, homophthalic anhydride 2-carboxyphenylacetic anhydride | |
4H-isochromene-1,3-dione |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H500
EINECSNumber : 211-873-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only