missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Holmium standard solution, for AAS, 1mg/mL Ho in 2% HNO3, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 196025000

 View more versions of this product

Catalog No. AC196025000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Holmium standard solution
7440-60-0
Solution
995 to 1005 μg/mL of Ho in 2% in 2% HNO3 (20°C)
MFCD00011049
Clear
1 mg/ml Ho in 2% HNO3
For AAS
Ho
164.93
Plastic bottle
500mL

Chemical Identifiers

7440-60-0
164.93
Holmium standard solution, 1 mg/ml Ho in 2% HNO3
Ho
MFCD00011049
For AAS

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning