Please login to your online account to display your discounted pricing

Holmium standard solution, for AAS, 1mg/mL Ho in 2% HNO3, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Holmium standard solution
Name Note 1 mg/ml Ho in 2% HNO3
CAS 7440-60-0
Purity Grade For AAS
Molecular Formula Ho
Formula Weight 164.93
MDL Number MFCD00011049
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC196025000

 
thermo fisher scientific
196025000
Plastic bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solution
Quantity 500mL
Color Clear
CAS 7440-60-0
Concentration 995 to 1005 μg/mL of Ho in 2% in 2% HNO3 (20°C)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Plastic bottle
Molecular Formula Ho
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Holmium standard solution
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00011049
Name Note 1 mg/ml Ho in 2% HNO3
Purity Grade For AAS
Formula Weight 164.93

For atomic adsorption spectroscopy

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.