Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium powder, -40 mesh, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Holmium powder
MDL Number MFCD00011049
UN Number UN1383
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA0014706

 
alfa aesar™
0014706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Holmium powder
TSCA TSCA
Physical Form Powder
Assay 99.9%
Assay Percent Notes (REO)
MDL Number MFCD00011049
UN Number UN1383
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H250-H260-H252
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
Health Hazard 3 GHS P Statement
P210-P222-P231+P232-P370+P378c-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Handle under inert gas.
Protect from moisture.
In case of fire: Use for
Storage Note 1
  • Packaged under argon
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC METALS, N.O.S.
Mesh Size −40 Mesh

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P210-P222-P231+P232-P370+P378c-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Handle under inert gas.
Protect from moisture.
In case of fire: Use for

GHS H Statement
H250-H260-H252
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.

  • Packaged under argon