Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium pieces, distilled dendritic, 99.9% (REO)

$84.53 - $84.53

Chemical Identifiers

Chemical Name or Material Holmium pieces
CAS 7440-60-0
Name Note Distilled dendritic
MDL Number MFCD00011049
UN Number UN3178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4151504

  SDS
alfa aesar™
4151504
2g Each for $84.53
Description & Specifications

Specifications

Quantity 2g
Health Hazard 3 GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99.9%
TSCA TSCA
MDL Number MFCD00011049
Odor Odorless
Assay Percent Notes (REO)
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
Physical Form Pieces
CAS 7440-60-0
Storage Note 1
  • Packaged under argon
UN Number UN3178
Health Hazard 2 GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Health Hazard 1 Danger
Color Dark gray to silver
Name Note Distilled dendritic
Chemical Name or Material Holmium pieces

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

  • Packaged under argon