Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium(III) sulfate hydrate, 99.99%

$83.93 - $326.97

Chemical Identifiers

Chemical Name or Material Holmium(III) sulfate hydrate
CAS 15622-40-9
Molecular Formula Ho2O12S3·xH2O
Linear Formula Ho2(SO4)3·xH2O
Formula Weight 618.03 (Anhydrous)
MDL Number MFCD00149740
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1709214

  SDS
alfa aesar™
A1709214
25g Each for $326.97

AAA1709206

  SDS
alfa aesar™
A1709206
5g Each for $83.93
Description & Specifications

Specifications

Formula Weight 618.03 (Anhydrous)
Chemical Name or Material Holmium(III) sulfate hydrate
TSCA TSCA
Assay 99.99%
Molecular Formula Ho2O12S3·xH2O
Linear Formula Ho2(SO4)3·xH2O
MDL Number MFCD00149740
CAS 15622-40-9
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.