missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Holmium(III) oxide, Beige powder or lump, REacton™, 99.99% (REO), Alfa Aesar™

$196.08 - $544.84

Chemical Identifiers

CAS 12055-62-8
Molecular Formula Ho2O3
MDL Number MFCD00011053
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1128109
View Documents
Thermo Fisher Scientific
1128109
10g
Each for $196.08
 
AA1128118
View Documents
Thermo Fisher Scientific
1128118
50g
Each for $544.84
 
Description

Description

Specifications

Chemical Identifiers

12055-62-8
Ho2O3
MFCD00011053

Specifications

12055-62-8
Ho2O3
Insoluble in water. Soluble in perchloric acids.
Powder
Beige
10g
99.99% (REO)
MFCD00011053
377.86
Odorless
2360°C
Holmium(III) oxide
Safety and Handling

Safety and Handling

EINECSNumber : 235-015-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS