missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Holmium(III) oxide, 99.995% (REO), Alfa Aesar™, Reacton™

Manufacturer:  Thermo Fisher Scientific 1091003

 View more versions of this product

Catalog No. AA1091003



Specifications

Specifications

Holmium(III) oxide
99.995% (REO)
MFCD00011053
377.86
8.36g/cm3
Beige
12055-62-8
Ho2O3
Beige powder or lump
Powder or Lump
Odorless
1g

Chemical Identifiers

12055-62-8
Ho2O3
MFCD00011053
Safety and Handling

Safety and Handling

EINECSNumber : 235-015-3

Documents