Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium(III) oxide, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Holmium(III) oxide
CAS 12055-62-8
Formula Weight 377.86
MDL Number MFCD00011053
Merck Index 14,4727
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3635709

  SDS
alfa aesar™
3635709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3635718

  SDS
alfa aesar™
3635718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 377.86
Chemical Name or Material Holmium(III) oxide
TSCA TSCA
Odor Odorless
Particle Size 3 to 12mm
Physical Form Sintered Lump
Assay 99.9%
Assay Percent Notes (REO)
MDL Number MFCD00011053
CAS 12055-62-8
Merck Index 14,4727
Melting Point 2360°C
Quantity 10g