missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium(III) nitrate pentahydrate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1458809

 View more versions of this product

Catalog No. AA1458809



Specifications

Specifications

Holmium(III) nitrate pentahydrate
(REO)
MFCD00149737
holmium nitrate pentahydrate, ho.3no3.5h2o, holmium iii nitrate hydrate, holmium nitrate, holmium nitrate-water 1/3/5, holmium 3+ pentahydrate trinitronate, holmium iii nitrate pentahydrate, holmium 3+ pentahydrate trinitrate, nitric acid,holmium 3+ salt, pentahydrate 8ci,9ci, holmium iii nitrate pentahydrate trace metals basis
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Ho+3]
441.017
441.02 (350.94 Anhydrous)
99.9%
Odorless
10g
14483-18-2
H10HoN3O14
UN1477
BJUGWWDCFYEYOA-UHFFFAOYSA-N
holmium(3+);trinitrate;pentahydrate
25021754
Crystalline Aggregates
Reacton™
Hygroscopic

Chemical Identifiers

14483-18-2
441.017
BJUGWWDCFYEYOA-UHFFFAOYSA-N
25021754
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Ho+3]
H10HoN3O14
MFCD00149737
holmium nitrate pentahydrate, ho.3no3.5h2o, holmium iii nitrate hydrate, holmium nitrate, holmium nitrate-water 1/3/5, holmium 3+ pentahydrate trinitronate, holmium iii nitrate pentahydrate, holmium 3+ pentahydrate trinitrate, nitric acid,holmium 3+ salt, pentahydrate 8ci,9ci, holmium iii nitrate pentahydrate trace metals basis
holmium(3+);trinitrate;pentahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents