missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium(III) iodide, ultra dry, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1363514

 View more versions of this product

Catalog No. AA1363514



Description

Description

  • Ampouled under argon
Specifications

Specifications

Holmium(III) iodide
13813-41-7
MFCD00049745
545.64
99.99%
Hygroscopic
Ultra dry
(REO)
14,4727
Flakes
Ampule
25g

Chemical Identifiers

13813-41-7
MFCD00049745
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Documents