Learn More
Alfa Aesar™ Holmium(III) iodide, ultra dry, 99.99% (REO)
Manufacturer: Alfa Aesar™ 1363506
Description
- Ampouled under argon
Specifications
Holmium(III) iodide | |
13813-41-7 | |
MFCD00049745 | |
545.64 | |
99.99% | |
Hygroscopic |
Ultra dry | |
(REO) | |
14,4727 | |
Flakes | |
Ampule | |
5g |
Chemical Identifiers
13813-41-7 | |
MFCD00049745 |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
TSCA : TSCA
storageNote1 :
- Ampouled under argon