missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium(III) fluoride, anhydrous, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1127814

 View more versions of this product

Catalog No. AA1127814



Specifications

Specifications

Holmium(III) fluoride
13760-78-6
MFCD00016142
Insoluble in water.
Crystalline Lump
Reacton™
Odorless
1143°C
Anhydrous
(REO)
UN3288
221.93
99.99%
7.6 g/mL
Hygroscopic
25g

Chemical Identifiers

13760-78-6
MFCD00016142
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 237-352-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents