missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium(III) chloride, ultra dry, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 4717806

 View more versions of this product

Catalog No. AA4717806



Description

Description

  • Ampouled under argon
Specifications

Specifications

Holmium(III) chloride
10138-62-2
Cl3Ho
14,4727
PYOOBRULIYNHJR-UHFFFAOYSA-K
trichloroholmium
24992
Beads
3.70g/mL
Odorless
Hygroscopic
5g
Ultra dry
(metals basis)
MFCD00011051
holmium chloride, holmium trichloride, holmium chloride hocl3, holmium iii chloride, wln: ho n-o3*3, holmium iii chloride, anhydrous, holmium chloride, holmium iii chloride, anhydrous, ?hocl3, holmium iii chloride, anhydrous, powder trace metals basis, 3ar,8ar---4,4,8,8-tetrakis 3,5-diethylphenyl tetrahydro-2,2-dimethyl-6-phenyl-1,3-dioxolo 4,5-e dioxaphosphepin
Cl[Ho](Cl)Cl
271.28
271.29
99.95%
−10 Mesh
Ampule
718°C

Chemical Identifiers

10138-62-2
271.28
PYOOBRULIYNHJR-UHFFFAOYSA-K
24992
Cl[Ho](Cl)Cl
Cl3Ho
MFCD00011051
holmium chloride, holmium trichloride, holmium chloride hocl3, holmium iii chloride, wln: ho n-o3*3, holmium iii chloride, anhydrous, holmium chloride, holmium iii chloride, anhydrous, ?hocl3, holmium iii chloride, anhydrous, powder trace metals basis, 3ar,8ar---4,4,8,8-tetrakis 3,5-diethylphenyl tetrahydro-2,2-dimethyl-6-phenyl-1,3-dioxolo 4,5-e dioxaphosphepin
trichloroholmium
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MS7350000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

SDS Documents