Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium(III) chloride hexahydrate, REacton™, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Holmium(III) chloride hexahydrate
CAS 14914-84-2
Purity Grade Reacton™
Linear Formula HoCl3·6H2O
Formula Weight 379.38 (271.29 Anhydrous)
MDL Number MFCD00149739
Merck Index 14,4727
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1127714

  SDS
alfa aesar™
1127714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1127722

  SDS
alfa aesar™
1127722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 379.38 (271.29 Anhydrous)
Purity Grade Reacton™
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Storage Note 1 2°C to 8°C
Physical Form Crystalline Aggregates
Health Hazard 1 Warning
CAS 14914-84-2
Linear Formula HoCl3·6H2O
Assay Percent Notes (REO)
Merck Index 14,4727
Odor Odorless
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00149739
Assay 99.9%
Melting Point 718°C
Sensitivity Hygroscopic
Quantity 25g
Chemical Name or Material Holmium(III) chloride hexahydrate
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2°C to 8°C