Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium(III) chloride, anhydrous, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Holmium(III) chloride
CAS 10138-62-2
Name Note Anhydrous
Formula Weight 271.29
MDL Number MFCD00011051
Merck Index 14,4727
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1868106

 
alfa aesar™
1868106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1868114

 
alfa aesar™
1868114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 271.29
Chemical Name or Material Holmium(III) chloride
TSCA TSCA
Mesh Size −20 Mesh
Odor Odorless
Name Note Anhydrous
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00011051
CAS 10138-62-2
Merck Index 14,4727
Melting Point 718°C
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 3.7g/mL
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.