Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium(III) carbonate hydrate, REacton™, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Holmium(III) carbonate hydrate
CAS 38245-34-0
Purity Grade Reacton™
Formula Weight 509.88 (Anhydrous)
MDL Number MFCD00011497
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1458909

 
alfa aesar™
1458909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1458918

 
alfa aesar™
1458918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 509.88 (Anhydrous)
Chemical Name or Material Holmium(III) carbonate hydrate
TSCA TSCA
Purity Grade Reacton™
Physical Form Powder
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
MDL Number MFCD00011497
CAS 38245-34-0
Quantity 10g