missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium(III) carbonate hydrate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1458918

 View more versions of this product

Catalog No. AA1458918



Specifications

Specifications

Holmium(III) carbonate hydrate
(REO)
MFCD00011497
XXHLSAONJAUANQ-UHFFFAOYSA-H
holmium(3+);tricarbonate;hydrate
57350480
Powder
Reacton™
50g
38245-34-0
C3H2Ho2O10
holmium carbonate hydrate, holmium iii carbonate hydrate, acmc-20akrc, holmium carbonate-water 2/3/1, holmium iii carbonate hydrate, reacton
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.[Ho+3].[Ho+3]
527.9
509.88 (Anhydrous)
99.9%
Hygroscopic

Chemical Identifiers

38245-34-0
527.9
XXHLSAONJAUANQ-UHFFFAOYSA-H
57350480
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.[Ho+3].[Ho+3]
C3H2Ho2O10
MFCD00011497
holmium carbonate hydrate, holmium iii carbonate hydrate, acmc-20akrc, holmium carbonate-water 2/3/1, holmium iii carbonate hydrate, reacton
holmium(3+);tricarbonate;hydrate
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents