missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium(III) bromide hydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1320318

 View more versions of this product

Catalog No. AA1320318



Specifications

Specifications

Holmium(III) bromide hydrate
(REO)
MFCD00209652
holmium iii bromide hydrate, holmium bromide hydrate, holmium bromide hobr3 , hydrate 9ci, holmium bromide hydrate
O.[Br-].[Br-].[Br-].[Ho+3]
422.657
404.66 (Anhydrous)
99.99%
Hygroscopic
50g
223911-98-6
Br3H2HoO
14,4727
ZSFPAAHZCBVDRA-UHFFFAOYSA-K
holmium(3+);tribromide;hydrate
57375929
Crystalline
Reacton™
914°C

Chemical Identifiers

223911-98-6
422.657
ZSFPAAHZCBVDRA-UHFFFAOYSA-K
57375929
O.[Br-].[Br-].[Br-].[Ho+3]
Br3H2HoO
MFCD00209652
holmium iii bromide hydrate, holmium bromide hydrate, holmium bromide hobr3 , hydrate 9ci, holmium bromide hydrate
holmium(3+);tribromide;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents