missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium(III) acetate hydrate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1459118

 View more versions of this product

Catalog No. AA1459118



Specifications

Specifications

Holmium(III) acetate hydrate
(REO)
holmium iii acetate hexahydrate, holmium iii acetate hydrate, reacton reo, holmium iii acetatehexahydrate, holmium acetate-water 1/3/6
Crystalline Aggregates
Reacton™
312619-49-1
MFCD00150648
342.07 (Anhydrous)
99.9%
50g

Chemical Identifiers

312619-49-1
MFCD00150648
holmium iii acetate hexahydrate, holmium iii acetate hydrate, reacton reo, holmium iii acetatehexahydrate, holmium acetate-water 1/3/6
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents