missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium foil, 1.0mm (0.04 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 10211FF

 View more versions of this product

Catalog No. AA10211FF



Description

Description

  • Packaged under argon
Specifications

Specifications

Holmium foil
25 x 25mm
7440-60-0
Mylar under Argon
Ho
acmc-1c9ym
[Ho]
164.93
CHEBI:49648
99.9%
≈5.5g/25x25mm
1.0mm (0.04 in.) thick
Odorless
(REO)
MFCD00011049
KJZYNXUDTRRSPN-UHFFFAOYSA-N
holmium
23988
Foil

Chemical Identifiers

7440-60-0
164.93
KJZYNXUDTRRSPN-UHFFFAOYSA-N
23988
holmium
Ho
MFCD00011049
acmc-1c9ym
CHEBI:49648
[Ho]
Safety and Handling

Safety and Handling

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents