Please login to your online account to display your discounted pricing

Alfa Aesar™ Holmium foil, 0.3mm (0.01 in.) thick, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Holmium foil
CAS 7440-60-0
Name Note 0.3mm (0.01 in.) thick
MDL Number MFCD00011049
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10210FF

  SDS
alfa aesar™
10210FF
25 x 25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10210FI

  SDS
alfa aesar™
10210FI
50 x 50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25 x 25mm
Packaging Mylar under Argon
TSCA TSCA
CAS 7440-60-0
Assay Percent Notes (REO)
Weight ≈1.65g/25x25mm
Name Note 0.3mm (0.01 in.) thick
Storage Note 1
  • Packaged under argon
MDL Number MFCD00011049
Assay 99.9%
Chemical Name or Material Holmium foil
Odor Odorless
Physical Form Foil

  • Packaged under argon

Safety and Handling

  • Packaged under argon