missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hoefer™ TE42 and TE62 Tank Blotting Units
Perform high-intensity protein transfers in one hour
Manufacturer: Hoefer™ TE42
Description
- Over 1.5m of platinum wire per electrode provides uniform field strength for efficient transfers
- Hinged, color-coded cassettes enable simple loading and easy orientation of gel sandwich
- Buffer volumes of 4 to 5L required, depending on number of cassettes processed
- Model TE62 offers a built-in heat exchanger in the base of the unit
- Model TE42 can be used with optional heat exchanger (Mfr. No. TE47)
Certifications
CE marked, CSA C22.2 1010.1, EN61010-1, UL3101-1
Specifications
2 A | |
Up to 2, 15 x 21 cm Gel (with cooling), up to 4, 15 x 21 cm Gel (without cooling) | |
Standard Transfer Tank | |
13 cm | |
Lower chamber, safety lid with high voltage Leads, 2 electrode panels, 2 gel cassettes, four 0.13 in. thick Foam sponges, two 0.25 in. thick Foam sponges, 25 sheets blotting Paper, cassette hook | |
30.5 cm | |
45°C | |
135.25 to 169.07 oz. | |
200 w | |
28 cm |
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE | |
12 x 11.02 x 5.11 in. (30.5 x 28 x 13 cm) | |
5.11 in. | |
Up to 80% | |
12 in. | |
113°F | |
100 V | |
4000 to 5000 mL | |
11.02 in. |