missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hoefer™ Submarine Gel Electrophoresis Units: SUB10 Mini-Plus Horizontal
Ideal for routine preparatory and analytical electrophoresis techniques
Manufacturer: Hoefer™ SUB10
Specifications
200 mA | |
9.05 x 6.4 in. (23 x 16.5 cm) | |
Routine preparatory and analytical electrophoresis technique | |
80% | |
9.05 in. | |
23 cm | |
80 | |
45°C | |
0.45 L | |
6.4 in. | |
16.5 cm |
EN61010-1, CE | |
Mini Plus Submarine Gel Electrophoresis Unit | |
6.5 cm | |
Buffer chamber assembly, safety lid with high voltage Leads, running tray with Silicone gasket, 1mm thick, 16-well combs, colored loading strips | |
3.9 in. | |
10 cm | |
113°F | |
300 V | |
60 w | |
4.5 in. | |
11.5 cm |