Please login to your online account to display your discounted pricing

Hoefer™ SUBHT High Throughput Submarine Gel Unit


Ideal for high throughput screening applications like single strand polymorphism (SSP) analysis following PCR

Manufacturer: hoefer™  SUBHT

 View more versions of this product

Catalog No. 03-500-401

  • / Each
 

Description & Specifications

Specifications

Height (Metric) 7cm
Length (English) 13.1 in.
Length (Metric) 33.5cm
Width (English) 8.46 in.
Width (Metric) 21.5cm
Amperage 200mA
Certifications/Compliance EN61010-1, CE
Product Type High throughput submarine gel unit
For Use With (Equipment) SUBHT high throughput submarine gel unit
Includes Buffer chamber assembly, safety lid with high voltage Leads, running tray with Silicone gasket, 1mm thick, 16-well combs, colored loading strips, Buffer recirculation ports
Length (English) Gel 12.9 in.
Length (Metric) Gel 33cm
Sample Size 120
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Voltage 300V
Width (English) Gel 8.4 in.
Volume (Fill) Metric 1.2L
Width (Metric) Gel 21.5cm
Dimensions (L x W) 13.1 x 8.46 in. (33.5 x 21.5cm)