missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hoefer™ SUBHT High Throughput Submarine Gel Unit

Ideal for high throughput screening applications like single strand polymorphism (SSP) analysis following PCR

Manufacturer:  Hoefer™ SUBSG

 View more versions of this product

Catalog No. 03-500-410



Includes:
  • Buffer chamber assembly
  • Safety lid, high-voltage leads
  • Fluorescent Labeled Running Tray with Casting Gates
  • Four 1mm thick 30-well combs
  • Colored Loading Strips
  • Two Buffer Recirculation Ports

Description

Description

  • Fast, error-free loading: load an entire 96-well plate using a multichannel pipetter
  • Simplified Analysis: Fluorescent labels for each sample printed on a UV transparent gel tray allows for instant sample identification (matched to a 96-well plate) during post-run visualization
  • High Throughput: Run up to 120 samples (96-well plate plus 24 control and/or marker samples) in 30 minutes
  • Better Reproducibility: Eliminate variability caused by running buffer pH drift and ionic gradient formation with optional buffer recirculation capability
  • Reduced Buffer Needs: Compact tank design needs less buffer to cover gel which not only reduces buffer consumption but provides greater control of voltage gradients and run times
  • Maximum Flexibility: Fully compatible with a wide range of combs and accessories to meet all your high throughput and standard gel analysis needs
  • This high throughput horizontal gel unit can accommodate up to 120 samples and resolve DNA strands from 50-1000 base pairs in 30 minutes
Specifications
  • Gel Dimensions: 15 x 15cm
  • Maximum Buffer Volume: 1.2L
  • Maximum Power Settings: 300V, 200mA, 60W
  • Maximum Sample Capacity: 120
  • Maximum Running Temperature: 45°C
  • Indoor Use: 4-40°C
  • Humidity: up to 80%
  • Unit Dimensions: 33.5L x 21.5W x 7cmH
  • Safety Certifications: EN61010-1, CE

Applications:

Single Strand Polymorphism (SSP) analysis

Specifications

Specifications

Silicone Gasket
SUBHT high throughput submarine gel unit
Silicon
Documents