Hoefer™ SUB15 Standard Submarine Combs
Accessories for SUB15 Standard Submarine
$190.50 - $198.67
Specifications
For Use With (Equipment) | SUB15 Standard Submarine |
---|---|
Type | Comb |
Depth (English) Pan | 0.19 in. |
Depth (Metric) Pan | 0.5 cm |
Catalog Number | Mfr. No. | Thickness (English) | Thickness (Metric) | No. of Wells | Width (Metric) Well | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Thickness (English) | Thickness (Metric) | No. of Wells | Width (Metric) Well | Price | Quantity | ||
03-500-761
|
Hoefer™
SUB15C1.516MC |
0.05 in. | 0.15 cm | 16 | 0.64 cm |
Each for $190.50
|
|
03-500-770
|
Hoefer™
SUB15C1.530MC |
0.05 in. | 0.15 cm | 30 | 0.25 cm |
Each for $190.50
|
|
03-500-760
|
Hoefer™
SUB15C116MC |
0.03 in. | 0.1 cm | 16 | 0.64 cm |
Each for $190.50
|
|
03-500-759
|
Hoefer™
SUB15C210 |
0.07 in. | 0.2 cm | 16 | 1.28 cm |
Each for $198.67
|
|
03-500-757
|
Hoefer™
SUB15C110 |
0.03 in. | 0.1 cm | 10 | 1.28 cm |
Each for $198.67
|
|
03-500-766
|
Hoefer™
SUB15C125 |
0.03 in. | 0.1 cm | 25 | 0.35 cm |
Each for $198.67
|
|
03-500-768
|
Hoefer™
SUB15C225 |
0.07 in. | 0.2 cm | 25 | 0.35 cm |
Each for $198.67
|
|
03-500-763
|
Hoefer™
SUB15C120 |
0.03 in. | 0.1 cm | 20 | 0.59 cm |
Each for $198.67
|
|
03-500-762
|
Hoefer™
SUB15C216MC |
0.07 in. | 0.2 cm | 20 | 0.64 cm |
Each for $198.67
|
|
03-500-769
|
Hoefer™
SUB15C130MC |
0.03 in. | 0.1 cm | 30 | 0.25 cm |
Each for $190.50
|
|
03-500-771
|
Hoefer™
SUB15C230MC |
0.07 in. | 0.2 cm | 30 | 0.25 cm |
Each for $190.50
|
|
Description
- Ranges from 10, 16, 20, 25 and 30 Wells
- 1, 1.5 and 2 mm Thickness
- 2.5 to 12.8 mm Well Width
- Min. 12, max. 128 μL Sample Volume
Specifications
SUB15 Standard Submarine | |
Comb | |
0.19 in. | |
0.5 cm |