Hoefer™ SG Series Gradient Makers
Ideal for generating gradients of polyacrylamide, sucrose, and cesium chloride
$980.22 - $1,419.81
Specifications
Type Gradient Maker
Includes Gradient maker w/PTFE Valves , Luer outlet fitting, support rod, adapter barbed fitting and 22G needle
View More Specs
Products 5
Catalog Number Mfr. No. For Use With (Equipment) Volume (English) Volume (Metric) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Volume (English) Volume (Metric) Price Quantity  
36-100-3970
View Documents Promotion Details
Hoefer™
SG100
Mighty Small™ Gel Caster SE 275 (for 4 Gels) or SE 215 (for 10 Gels) 3.38 oz. 100 mL
Each for $1,321.81
36-100-3967
View Documents Promotion Details
Hoefer™
SG50
Mighty Small™ Gel Caster SE 275 (for 4 Gels) or SE 215 (for 10 Gels) 1.69 oz. 50 mL
Each for $1,193.17
36-100-3968
View Documents Promotion Details
Hoefer™
SG30
Mighty Small™ Gel Caster SE 275 (for 4 Gels) or SE 215 (for 10 Gels) 1 oz. 30 mL
Each for $997.44
36-100-3966
View Documents Promotion Details
Hoefer™
SG500
SE 615, SE 915 Multiple Gel Casters 16.9 oz. 500 mL
Each for $1,419.81
36-100-3969
View Documents Promotion Details
Hoefer™
SG15
Mighty Small™ Gel Caster SE 275 (for 4 Gels) or SE 215 (for 10 Gels) 0.5 oz. 15 mL
Each for $980.22
Description
Description
Models SG 15, SG 30, SG 50, and SG 100
  • Milled of heavy acrylic plastic with high-quality, leak-free PTFE-resin valves
  • Offer a flat base for stability on magnetic stirrers; support rod can be attached to ring stand for additional stability
  • Accept Luer or 4 mm ID tubing; with included adapters can also accept 2 mm ID or 22-gauge tubing
  • SG 100 is ideal for the Hoefer Mighty Small™ Gel Casters SE 275 (for four gels) or SE 215 (for 10 gels)
Model SG 500
  • Made of two cylindrical acrylic chambers joined and mounted on a flat acrylic base
  • Uses a simple push-pull valve to open and close the passage between the reservoir and mixing chambers
  • Suitable for forming polyacrylamide gradients or buffer gradients used with medium to small chromatography columns
  • Ideal for use with the Hoefer SE 615 or SE 915 Multiple Gel Casters
Specifications
Specifications
Gradient Maker
Gradient maker w/PTFE Valves , Luer outlet fitting, support rod, adapter barbed fitting and 22G needle