Please login to your online account to display your discounted pricing
Hoefer™ SE640 Wide-Mini Vertical Electrophoresis Unit
Wide-mini system, ideal for rapid screening of multiple samples
$1,977.95 - $2,368.40
Products
Catalog Number Mfr. No. Price Quantity    
36-101-9553
hoefer™
SE640151.5
Each for $2,368.40
SE640
hoefer™
SE640
Each for $1,977.95
Description & Specifications
The Hoefer Wide-Mini Vertical Electrophoresis unit offers extra wide format ideal for quick, high throughput runs. With accessory divider plate you can complete a run of up to 112 samples in as little as 40 minutes.
  • Maximum glass plate size 18 x 8cm, permits rapid screening of up to 112 samples at one time
  • Design permits gels to be surrounded uniformly by buffer so heat dissipation is efficient; bands are sharp and lanes are straight
  • Accommodates denaturing or native polyacrylamide gels
  • Run up to four second dimension gels suing IEF strips or tube gels
  • Ideal for optimizing 2DE protocols
  • Storage temperature: +15°C to 30°C
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.