Please login to your online account to display your discounted pricing

Hoefer™ SE640 Wide-Mini Vertical Electrophoresis Unit


Wide-mini system, ideal for rapid screening of multiple samples

Manufacturer: hoefer™  SE640

 View more versions of this product

Catalog No. SE640

  • / Each
 

Description & Specifications

Specifications

Temperature (English) Maximum 113°F
Depth (English) 5.51 in.
Temperature (Metric) Storage 15° to 30°C
Wattage 50w
Width (English) Exterior 12.59 in.
Width (Metric) Gel 18cm
Height (English) Exterior 11.41 in.
Height (Metric) Exterior 29cm
Voltage 1000V
Includes Lower buffer chamber (tank), safety lid with leads, upper buffer chamber with cathode and anode electrode fin, 3 sets of glass plates (18 x 8cm), four 8cm clamp assemblies, 6 cams, dual gel casting stand with leveling base and level, buffer dam, spacer-mate alignment template, wonder wedge plate separation tool and gel seal, bubble level
Height (English) Gel 3.14 in.
Temperature (Metric) Maximum 45°C
Depth (Metric) 14cm
Humidity Range Up to 80%
Amperage 500mA
Certifications/Compliance EN61010-1, CSA C22.2 1010.1, CE
Width (English) Gel 7.08 in.
Width (Metric) Exterior 32cm