Please login to your online account to display your discounted pricing

Hoefer™ SE 900 Large Format Vertical Electrophoresis System

Ideal for use with 24cm IPG strips in the second dimension of 2-D electrophoresis

Specifications

Amperage 1000mA
Certifications/Compliance EN61010-1, UL61010-1, CSA C22.2, CE
Depth (English) 7.87 in.
Depth (Metric) 20cm
Height (English) Exterior 16.92 in.
View More Specs
Includes:  Buffer chamber with pumps, safety lid with leads, gel rack, 10 ft. tubing, two pair recirculation bath connectors, drain connector, seven tubing clamps, gel seal, wonder wedge plate separation tool
Mfr. No. SE900-1.0 also includes: Multicaster with ported face plate, six red spring clamps, two thumb screws, ten divider sheets, filler plug, cassette blank, gel seal, six glass hinge casettes
Products
Catalog Number Mfr. No. Includes Price Quantity    

SE90010NA

 
hoefer™
SE9001.0NA
Separation tank, tubing kit, lid with high voltage Leads, multiple gel caster, internal PAGE rack, hinged Glass cassettes CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Amperage 1000mA
Certifications/Compliance EN61010-1, UL61010-1, CSA C22.2, CE
Depth (English) 7.87 in.
Depth (Metric) 20cm
Height (English) Exterior 16.92 in.
Height (Metric) Exterior 43cm
Width (English) Exterior 16.92 in.
Width (English) Gel 9.84 in.
Width (Metric) Exterior 43cm
Width (Metric) Gel 25cm
Height (English) Gel 7.87 in.
Humidity Range Up to 80%
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Temperature (Metric) Storage 15° to 30°C
Voltage 600V
Wattage 150w
Weight (English) 29.98 lb.
Weight (Metric) 13.6kg

  • Runs 1-6 gels
  • Uses hinged cassettes for convenience and straight dye fronts
  • Easy to use — no clamps, no loose parts, bottom drain port
  • Utilizes internal pump for even temperature distribution
  • Compatible with a recirculating chiller for active cooling

UL61010-1, CSAC22.2 No. 61010-1, EN61010-1:2001, CE Marked

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.