Hoefer™ Plate and Spacers for SE 660 Vertical Electrophoresis Unit
For use with SE660, SE600X Chroma, SE600, SE640, SE410 and SE400
Supplier: Hoefer™ SE6602D
Description
- Ranges from 18 x 16 to 18 x 24 cm
- 2 pcs
- Notched
Specifications
SE 660 Vertical Electrophoresis Unit | |
Notched Glass Plate Divider | |
Glass |