Please login to your online account to display your discounted pricing
Hoefer™ Mini and Standard Electrophoresis Blotting Systems
Economical equipment packages provide proven electrophoresis solutions for Western blotting
Manufacturer: hoefer™  360773
 View more versions of this product
Catalog No. 03-500-807
  • / Each
Description & Specifications
Specifications
Product Type Standard Electrophoresis Blotting System
Includes Deluxe dual cooled vertical unit, large semi dry transfer unit with built-in power supply
Amperage 500mA
Voltage 300V
Wattage 90w