Hoefer™ Mini and Standard Electrophoresis Blotting Systems
Economical equipment packages provide proven electrophoresis solutions for Western blotting
Manufacturer:  Hoefer™ 326223
 View more versions of this product
Catalog No. 36-102-1085
Description
Description
  • Comes with PS300B 300 V, 500 mA, 90 W Power Supply for all units
  • Save time and money with a combination package
Specifications
Specifications
Mini Electrophoresis Blotting System
300 V
500 mA
Mighty Small™ II mini deluxe vertical unit, Mighty Small™ mini transfer tank, power supply
90 w