Hoefer™ Mini and Standard Electrophoresis Blotting Systems
Economical equipment packages provide proven electrophoresis solutions for Western blotting
Manufacturer: Hoefer™ 326703
Description
- Comes with PS300B 300 V, 500 mA, 90 W Power Supply for all units
- Save time and money with a combination package
Specifications
Mini Electrophoresis Blotting System | |
300 V | |
500 mA |
Mighty Small™ II mini deluxe vertical unit, semi dry transfer unit with built-in power supply | |
90 w |