Please login to your online account to display your discounted pricing
Hoefer™ Mini-Peristaltic Pump
For use with horizontal units with buffer recirculation ports
$239.87 - $3,360.64
Specifications
For Use With (Equipment) Horizontal unit with Buffer recirculation port
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
03-500-412
hoefer™
PP24TB
Pump Head Tubing Each for $239.87
03-500-411
hoefer™
PP24
Mini Peristaltic Pump Each for $3,360.64
Description & Specifications
Specifications
For Use With (Equipment) Horizontal unit with Buffer recirculation port
The Mini-Peristaltic Pump can be used with either a single tube or two tubes simultaneously. Two of the Pump Head Tubing Pieces are included with the pump. Additional Pump Head Tubing Pieces may be purchased separately.
  • Two front panel controls provide flow rates from approximately 0.8 to 24.5mL/min.
  • Control knob provides variable adjustment from 0 to 100% of the selected flow rate range
  • Second control is a two position toggle switch marked x1, x2 which selects low or high flow rates
  • Four roller pump head is on top of the stout metal box
  • Back of the pump head rotates into an“open”position and either one or two tubes can be dropped into slots
  • Loaded section rotates back against spring loaded jaws and locks into place
  • Tubing is automatically in proper wiping contact with the pump head rollers
  • Each pump is provided with a 12.5mm (0.5 in.) rod clamp on the back so that multiple pumps can be mounted vertically on a lattice rod
CE marked